EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
alpha 2-HS glycoprotein/fetuin, a transforming growth factor-beta/bone morphogenetic protein antagonist, regulates postnatal bone growth and remodeling.
PMID 11901155
Abstract
Soluble transforming growth factor-beta (TGF-beta)/bone morphogenetic protein (BMP)-binding proteins are widely distributed in mammalian tissues and control cytokine access to membrane signaling receptors. The serum and bone-resident glycoprotein alpha2-HS-glycoprotein/fetuin (ASHG) binds to TGF-beta/BMP cytokines and blocks TGF-beta1 binding to cell surface receptors. Therefore, we examined bone growth and remodeling phenotypes in ASHG-deficient mice. The skeletal structure of Ahsg(-/-) mice appeared normal at birth, but abnormalities were observed in adult Ahsg(-/-) mice. Maturation of growth plate chondrocytes was impaired, and femurs lengthened more slowly between 3 and 18 months of age in Ahsg(-/-) mice. However, bone formation was increased in Ahsg(-/-) mice as indicated by greater cortical thickness, accelerated trabecular bone remodeling, and increased osteoblast numbers on bone surfaces. The normal age-related increase in cortical thickness and bone mineral density was accelerated in Ahsg(-/-) mice and was associated with increased energy required to fracture. Bone formation in response to implanted BMP cytokine extended further from the implant in Ahsg(-/-) compared with Ahsg(+/+) mice, confirming the interaction between ASHG and TGF-beta/BMP cytokines in vivo. Our results demonstrate that ASHG blocks TGF-beta-dependent signaling in osteoblastic cells, and mice lacking ASHG display growth plate defects, increased bone formation with age, and enhanced cytokine-dependent osteogenesis.
Related Materials