EMAIL THIS PAGE TO A FRIEND
The Journal of trauma
The use of chemical incapacitant sprays: a review.
PMID 11901348
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
77460
2-Chloroacetophenone, purum, ≥98.0% (GC)
C8H7ClO
C19686
2-Chloroacetophenone, 98%
C8H7ClO