EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Three-component cycloadditions: the first transition metal-catalyzed [5+2+1] cycloaddition reactions.
PMID 11902870
Abstract
Prompted by our studies of transition metal-catalyzed [4+4], [4+2], [5+2], and [6+2] cycloadditions and by the view that these two-component reactions could be intercepted by a third component of one or more atoms, a new three-component transition metal-catalyzed cycloaddition is described. This new [5+2+1] cycloaddition proceeds in good to excellent yield and with high or complete regioselectivity with a variety of carbonyl-substituted alkynes to give bicyclo[3.3.0]octenone adducts, resulting from transannular closure of the intermediate eight-membered-ring cycloadduct. Effects of concentration, temperature, pressure, and catalyst loading on the efficiency of the reaction are discussed. This process provides access to complex building blocks for synthesis based on simple, readily available components.