EMAIL THIS PAGE TO A FRIEND
The Lancet. Oncology
Supercomputer-designed drug protects against chemotherapy toxicity.
PMID 11905631
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000317 Mesna impurity D, European Pharmacopoeia (EP) Reference Standard
C4H10O6S4