EMAIL THIS PAGE TO A FRIEND
The journal of peptide research : official journal of the American Peptide Society
Topography of the neurotensin (NT)(8-9) binding site of human NT receptor-1 probed with NT(8-13) analogs.
PMID 11906607
Abstract
A series of neurotensin (NT)(8-13) analogs featuring substitution of the Arg8 and/or Arg9 residues with non-natural cationic amino acids was synthesized and evaluated for binding to the human NT receptor-1 (hNTR-1). The modifications were designed to probe specific steric and electrostatic requirements in the N-terminal cationic region of NT(8-13) for receptor binding as a general evaluation of the feasibility of incorporating minor structural changes into a peptide at a crucial polar receptor binding site. Many of the non-natural amino acids are more or less isosteric to Arg but more lipophilic as a result of addition of alkyl groups or through removal or replacement of NH character with methylene or methyl substituents, whereas others vary the distance between the cation and the alpha-amino acid carbon. Substitution of Arg8 with N(G)-alkylated Arg derivatives or homolysine (Hlys) maintained the subnanomolar affinity of NT(8-13) to the hNTR-1. Position 8 incorporation of Hlys produced the most favorable primary amine side-chain substitution to date. Moderate losses in affinity observed with position 9 substitutions were attributed to adverse steric effects. Doubly substituted [Hlys8, DAB9]NT(8-13), in which DAB is 2,4-diaminobutyric acid, was also prepared and tested as the shorter side-chain of DAB is known to be favored in position 9 of NT(8-13). This analog maintained 60% of NT(8-13) binding affinity making it the most favored des-guanidinium-containing analog known. These results demonstrate that adequate receptor binding affinity can be maintained over a structural range of Arg analogs, thus providing a range of peptides expected to exhibit altered pharmacokinetic properties. From the standpoint of the hNTR-1 cationic binding sites, these results help to map out the structural stringency inherent in the formation of a tight binding complex with NT(8-13) and related analogs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N5266
Neurotensin Fragment 8-13 acetate salt, ≥97% (HPLC)
C38H64N12O8