EMAIL THIS PAGE TO A FRIEND
Progress in molecular and subcellular biology
Proteasome inhibitors.
PMID 11908069
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D7910
3,4-Dichloroisocoumarin, serine protease inhibitor
C9H4Cl2O2