EMAIL THIS PAGE TO A FRIEND
Archives of toxicology
Covalent binding of nitrogen mustards to the cysteine-34 residue in human serum albumin.
PMID 11914777
Abstract
Covalent binding of various clinically important nitrogen mustards to the cysteine-34 residue of human serum albumin, in vitro and in vivo, is demonstrated. A rapid method for detection of these adducts is presented, based on liquid chromatography-tandem mass spectrometry analysis of the adducted tripeptide Cys*-Pro-Phe after digestion of the protein with Pronase.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B38503
Bis(2-chloroethyl)amine hydrochloride, 98%
C4H9Cl2N · HCl