EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
Allergic contact dermatitis from unsaturated polyester resin consisting of maleic anhydride, phthalic anhydride, ethylene glycol and dicyclopentadiene.
PMID 11918596
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
454338
Dicyclopentadiene, contains BHT as stabilizer
C10H12