EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Studies on novel peptidomimetics having bi-directional dispositions of hydroxylated D-Pro-Gly motifs anchored on a C(2)-symmetric iminosugar-based foundation.
PMID 11925214
Abstract
A rigid pyrrolidine based scaffold comprising of 2,5-dideoxy-2,5-imino-D-idaric acid (1) is developed. Attachment of peptide strands to the carboxylic groups at both ends of this novel template led to the peptidomimetics 2 and 3. Conformational analysis by NMR studies revealed that compounds 2b, 3b and 2c, 3c take interesting turn structures (C(2) symmetric for 2c and 3c) in DMSO-d(6) consisting of identical intramolecular hydrogen bonds at two ends between LeuNH --> sugar-OH as depicted in structure A, whereas 2a and 3a display structures with regular beta-turns with hydrogen bonds between LeuNH --> Boc-C=O in one-half of their molecular frameworks (structure B), characteristic of the turn structures commonly observed in "D-Pro-Gly"-containing peptides. These results suggest that a cis hydroxyl group at the 3-position of the proline residue favors a pseudo beta-turn-like nine-membered ring structure in hydroxyproline-containing peptides involving an intramolecular hydrogen bond between the hydroxyl and the i + 2 backbone amide.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P0880
Pro-Gly
C7H12N2O3