EMAIL THIS PAGE TO A FRIEND
Environmental pollution (Barking, Essex : 1987)
Evidence of differences in the biotransformation of organic contaminants in three species of freshwater invertebrates.
PMID 11926182
Abstract
Acute static bioassays were performed using three freshwater invertebrate species (the oligochaete Lumbriculus variegatus, the fingernail clam Sphaerium corneum and the larvae Chironomus riparius) exposed separately to a variety of 14C radiolabelled contaminants. The aim of this work was to investigate if the chemicals remained as parent compounds after the treatments. Chemicals used were 2,4-dichlorophenol; 2,4,5-trichlorophenol; pentachlorophenol; pyrene; Fenpropidin, and Trifluralin. Homogenates of the whole body tissue of each organism were prepared and total radioactivity was measured. Contaminants were then extracted into organic solvents and analysed by high-pressure liquid chromatography techniques. Chromatograms showed that most of the substances extracted were present as parent compounds in S. corneum and in L. variegatus. In contrast, for C. riparius a low proportion of the chemicals was recovered as parent compounds. These results suggest that different metabolic processes could take place in the different species.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46017
Fenpropidin, PESTANAL®, analytical standard
C19H31N