EMAIL THIS PAGE TO A FRIEND
Nihon eiseigaku zasshi. Japanese journal of hygiene
[Epidemiological studies on the etiology of biliary tract cancers].
PMID 11928319
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
04287
Chlornitrofen, analytical standard
C12H6Cl3N1O3