EMAIL THIS PAGE TO A FRIEND
The Journal of clinical endocrinology and metabolism
Overexpression of cyclin D1 and underexpression of p27 predict lymph node metastases in papillary thyroid carcinoma.
PMID 11932323
Abstract
Lymph node metastasis in papillary thyroid carcinoma increases the morbidity of treatment and the risk of local regional relapse and may also affect cure rates and survival. Factors that predict lymph node metastasis are, however, unclear. We analyzed 125 patients with papillary thyroid carcinoma for factors that predict lymph node metastasis. On univariate analysis, age, extrathyroidal extension, tumor focality, overexpression of cyclin D1, and underexpression of p27 predicted lymph node metastasis, whereas patient gender and tumor size did not. On multivariate analysis, extrathyroidal extension, overexpression of cyclin D1, and underexpression of p27 proved to be strong independent predictors of lymph node metastasis. We suggest that immunohistochemistry for cyclin D1 and p27 will prove valuable in identifying papillary thyroid carcinomas with metastatic potential.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
427M-9
p27Kip1 (SX53G8) Mouse Monoclonal Antibody