EMAIL THIS PAGE TO A FRIEND
Phytotherapy research : PTR
Chemical composition and biological activity of the volatiles of Anthemis melampodina and Pluchea dioscoridis.
PMID 11933125
Abstract
The volatile fractions obtained by hydrodistillation of the fresh leaves of Anthemis melampodina and Pluchea dioscoridis were analysed by GC-MS technique. Of 38 components identified in the volatile oil of A. melampodina, santolinatriene was the major component (27.33%). The oil was characterized by a high percentage of monoterpene hydrocarbons (49.94%) while sesquiterpene hydrocarbons and oxygenated sesquiterpenes represented only 7.41% and 11.43% of the oil. 36 components were identified in the volatile oil of P. dioscoridis. Farnesol was the major component (16.50%) accompanied by a high percentage of sesquiterpene alcohols. Oxygenated sesquiterpenes (26.43%) and sesquiterpene hydrocarbons (39.43%) represented the main constituents in the oil. P. dioscoridis showed a marked mosquito larvicidal activity against Culex pipiens (LC(50) 71.86 ppm), while A. melampodina was moderately active (LC(50) 139.42 ppm).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46037
Cyphenothrin, PESTANAL®, analytical standard
C24H25NO3