EMAIL THIS PAGE TO A FRIEND
Human mutation
An update on conformation sensitive gel electrophoresis.
PMID 11933188
Abstract
Conformation-sensitive gel electrophoresis (CSGE) was developed as a method of heteroduplex analysis to screen large multi-exon genes for sequence variation. The novelty of the method was in the use of a non-proprietary acrylamide gel matrix that used 1,4-bis (acrolyl) piperazine (BAP) as a cross linker with ethylene glycol and formamide as mildly denaturing solvents. The denaturing environment enhances the conformation polymorphism present in DNA heteroduplexes containing variations as small as single nucleotide polymorphisms (SNPs). CSGE has also been adapted for use on a fluorescent platform (F-CSGE) that resulted in higher throughput and sensitivity. Variation in sensitivity of CSGE has been studied extensively. The results demonstrate that the nature of the mismatched base in a defined sequence context has the most profound effect on the conformation of the heteroduplex. Additionally, the size of the PCR product, as well as the location of the mismatch within the PCR product, are two important parameters that determine the resolution of the mismatch-containing heteroduplexes during CSGE. Like any other mutation scanning technique, CSGE can have limited resolution of two closely linked sequence variations. For specific genes, like BRCA1 and BRCA2 where multiple SNPs are present in the coding sequence, each CSGE shift has to be sequenced to define the exact nature of the sequence change. In conclusion, CSGE scanning provides a powerful, cost-efficient way to scan genes with high sensitivity and specificity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D1538
Piperazine diacrylamide, For acrylamide gel electrophoresis
C10H14N2O2