EMAIL THIS PAGE TO A FRIEND
Natural product letters
1,5-dihydroxyanthraquinones and an anthrone from roots of Rumex crispus.
PMID 11942685
Abstract
From the roots of Rumex crispus, two known anthraquinones and a new one together with a new anthrone were isolated and the structures of compounds 1-4 were elucidated by spectroscopic means. The singlet oxygen generation capacity was tested with 1,3-diphenylisobenzofuran (DPBF) for compounds 1-4.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
105481
1,3-Diphenylisobenzofuran, 97%
C20H14O