EMAIL THIS PAGE TO A FRIEND
Journal of clinical laboratory analysis
Analysis of 3-hydroxydodecanedioic acid for studies of fatty acid metabolic disorders: preparation of stable isotope standards.
PMID 11948802
Abstract
Current diagnostic tests to detect disorders of fatty acids metabolism, such as long-chain hydroxyacyl CoA dehydrogenase deficiency (LCHAD), are hampered by insensitivity or a long delay time required for results. Children with LCHAD deficiency are known to excrete 3-hydroxydicarboxylic acids with chain lengths of 10-16 carbons, but a quantitative method to measure excretion of these potentially diagnostically important compounds has not been reported. We report synthetic schemes for synthesis of 3-hydroxydodecanedioic acid and a di-deuterated analog, suitable for use in a stable-isotope dilution mass spectrometric analytical approach. Evaluation of several common derivatization protocols to produce a volatile derivative for gas chromatography determined that trimethylsyl derivatives produced the best efficiency and stability. Positive-ion chemical ionization mass spectrometry provided the greatest yield of characteristic ions. These results indicate the basic reagents needed to develop sensitive and accurate 3-hydroxydodecanedioic acid measurements for diagnosis of LCHAD deficiency and other fatty acid oxidation disorders.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H3398
DL-β-Hydroxylauric acid, ≥99% (GC)
C12H24O3