EMAIL THIS PAGE TO A FRIEND
Biochemistry
The GTP-dependent restriction enzyme McrBC from Escherichia coli forms high-molecular mass complexes with DNA and produces a cleavage pattern with a characteristic 10-base pair repeat.
PMID 11955074
Abstract
The GTP-dependent restriction enzyme McrBC consists of two polypeptides: one (McrB) that is responsible for GTP binding and hydrolysis as well as DNA binding and another (McrC) that is responsible for DNA cleavage. It recognizes two methylated or hemimethylated RC sites (R(m)C) at a distance of approximately 30 to more than 2000 base pairs and cleaves the DNA close to one of the two R(m)C sites. This process is strictly coupled to GTP hydrolysis and involves the formation of high-molecular mass complexes. We show here using footprinting techniques, surface plasmon resonance, and scanning force microscopy experiments that in the absence of McrC, McrB binds to a single R(m)C site. If a second R(m)C site is present on the DNA, it is occupied independently by McrB. Whereas the DNA-binding domain of McrB forms 1:1 complexes with each R(m)C site and shows a clear footprint on both R(m)C sites, full-length McrB forms complexes with a stoichiometry of at least 4:1 at each R(m)C site, resulting in a slightly more extended footprint. In the presence of McrC, McrB forms high-molecular mass complexes of unknown stoichiometry, which are considerably larger than the complexes formed with McrB alone. In these complexes and when GTP is present, the DNA is cleaved next to one of the R(m)C sites at distances differing by one to five helical turns, suggesting that in the McrBC-DNA complex only a few topologically well-defined phosphodiester bonds of the DNA are accessible for the nucleolytic center of McrC.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I6875
5-Iodocytosine
C4H4IN3O