EMAIL THIS PAGE TO A FRIEND
Combinatorial chemistry & high throughput screening
Optimisation of polystyrene resin-supported Pt catalysts in room temperature, solvent-less, oct-l-ene hydrosilylation using methyldichlorosilane.
PMID 11966428
Abstract
Six precursor resins with systematic variation of porous parameters were prepared by suspension polymerisation using specific compositions of divinylbenzene, styrene vinylbenzyl chloride (VBC) and 2-ethylhexan-l-ol (a porogen). Surface areas from N(2) sorption and BET analysis were approximately 2-170 m(2)g-(1). The VBC content in each case was 38 mol% and these groups were aminated using the sodium salt of trimethylethylene diamine. Pt was introduced onto each resin at three different loadings (approximately 0.03, approximately 0.2 and approximately 0.4 mmol g-(1)) by appropriate manipulation of K(2)PtCl(6). The matrix of 18 resin-supported Pt complexes was then assessed for catalytic activity in the room temperature, solvent-less, hydrosilylation of oct-l-ene using methyldichlorosilane such that alkene: silane: Pt ratio was fixed at 2:1:1x10(-3). Though all the catalysts showed activity lower than that of homogeneous Speier s catalyst, most were sufficiently active to be potentially valuable heterogeneous catalysts in the laboratory, and indeed the plant. The most lightly loaded resins proved to be the least active. The remainder were recycled 5 times, and the best performers, the most highly loaded species, a further 5 times making 10 consecutive uses in all. A strong dependence on the porous structure of the resins was demonstrated with the activity rising systemically with the surface area. The two highest surface area highest loaded species displayed good activity even when used for the tenth time. The level of concurrent alkene isomerisation observed was very low throughout (<1%) making these heterogeneous species very selective as well as highly active. Overall the derived catalysts are excellent candidates for use in the research laboratory, and with further development could also be valuable in continuous processes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O4806
1-Octene, 98%
C8H16
74900
1-Octene, analytical standard
C8H16
W429300
1-Octene, 98%
C8H16