EMAIL THIS PAGE TO A FRIEND
Rapid communications in mass spectrometry : RCM
Electrospray ionization mass spectrometric studies of phosphorus oxychloride directed synthesis of homo-oligopeptide ester libraries.
PMID 11968121
Abstract
With the assistance of phosphorus oxychloride, alpha-amino acids were assembled into homo-peptides, which were analyzed by electrospray ionization mass spectrometry (ESI-MS) and multistage electrospray ionization mass spectrometry (ESI-MS/MS). On quenching with water or various alcohols, the reaction mixtures yielded the corresponding peptides or peptide esters, respectively. This paper reports a simple method to synthesize the homo-oligo-peptide-ester conjugated library by phosphorus oxychloride activation.
Related Materials