EMAIL THIS PAGE TO A FRIEND
Archives of pediatrics & adolescent medicine
Starting dose of levothyroxine for the treatment of congenital hypothyroidism: a systematic review.
PMID 11980555
Abstract
To determine the effect of levothyroxine sodium starting dose on cognitive development, growth, or behavior in children with congenital hypothyroidism identified by neonatal screening. Systematic review of cohort studies. Two analyses were performed: a between-study comparison of mean starting dose with mean developmental score and an analysis of the within-study effects of starting dose on cognitive development, growth, or behavior. The between-study comparison (14 cohort studies based on 1321 patients) found that the standardized mean IQ or developmental quotient scores ranged from 90 to 115 but were not associated with the mean starting dose of levothyroxine (P =.48). The within-study comparison of 4 cohort studies (based on 558 patients) that reported the effect of the starting dose of levothyroxine on cognitive development found no consistent effects. There was weak evidence for an effect of starting dose on growth (1 study) and on behavior problems (1 study). The evidence for an effect of starting dose of levothyroxine on cognitive development, growth, or behavior is too weak to justify recommendations in favor of high- or standard-dose regimens. More reliable information, based on a randomized controlled trial of starting dose or a meta-analysis of the individual patient data currently available, is required to inform treatment policies.