EMAIL THIS PAGE TO A FRIEND
MCN. The American journal of maternal child nursing
Sterile speculum exams, nitrazine, and ferning.
PMID 11984281
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
200182
Nitrazine Yellow, indicator grade, Dye content 85 %
C16H8N4Na2O11S2