EMAIL THIS PAGE TO A FRIEND
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry
Fluorometric determination of N-nitroso-N-methylurea with nicotinamide and acetophenone.
PMID 11990612
Abstract
A fluorometric method for the determination of N-nitroso-N-methylurea (NMU) has been developed. It is based on the N-methylation reaction of nicotinamide with NMU and a subsequent condensation reaction with acetophenone, followed by an acid treatment to form a fluorescent 2,7-naphthyridine derivative. This method enabled the determination of NMU in the range 0.05 - 2 nmol/200 microl with a relative standard deviation of ca. 3%. It was applied to the determination of NMU formed from a precursor N-methylurea (MU) under simulated gastric conditions containing nitrite and thiocyanate ions at pH 3.0 in the presence of fresh orange juice and milk. NMU was extracted by an Extrelut 20 column and then determined. The mean recoveries of NMU added to the simulated gastric juice containing water, orange juice and milk were 86.5, 85.1 and 69.8%, respectively. The amounts of NMU formed from MU were found to decrease to below 25% in the presence of orange juice and milk.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M86804
N-Methylurea, 97%
C2H6N2O