EMAIL THIS PAGE TO A FRIEND
Journal of Asian natural products research
New beyerane and isopimarane diterpenoids from Rhizophora mucronata.
PMID 11991193
Abstract
Chemical examination of the ethyl acetate extract of the roots of Rhizophora mucronata collected from Mangalore Coast resulted in the isolation of three more new diterpenoids, rhizophorins C-E (1-3) in addition to the two, rhizophorin A, (6R,11S,13S)-6,11,13-trihydroxy-2,3-seco-14-labden-2,8-olid-3-oic acid, and rhizophorin B, ent-3beta,20-epoxy-3alpha,18-dihydroxy-15-beyerene, recently reported. The structures of the new diterpenoids have been elucidated by a study of their physical and spectral data as 17-hydroxybeyer-15-en-3-one (1), 3beta,17-diacetoxy-15-beyeren-2-one (2) and 3beta,6alpha-diacetoxy-8(14),15-isopimaradien-2-one (3).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I6783 Isopimaric Acid, ≥98% (GC), powder
C20H30O2