EMAIL THIS PAGE TO A FRIEND
Journal of the history of medicine and allied sciences
Women's trials: the approval of the first oral contraceptive pill in the United States and Great Britain.
PMID 11995593
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
855871
Mestranol, 99%
C21H26O2