EMAIL THIS PAGE TO A FRIEND
Die Pharmazie
Nonaqueous capillary electrophoretic analysis of hexetidine in a commercial liquid formulation.
PMID 11998443
Abstract
A non-aqueous capillary electrophoresis method has been proposed to analyse hexetidine and its degradation products in a commercial liquid pharmaceutical formulation. The separation was achieved using a 37 cm fused silica capillary with a 75 microns internal diameter, an electrolyte of 120 mM sodium acetate buffer in methanol, at 16 kV and 25 degrees C. The results show that the proposed method is able to resolve hexetidine from its degradation products and/or impurities.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
259187
Hexetidine, mixture of stereoisomers
C21H45N3