EMAIL THIS PAGE TO A FRIEND
Mycopathologia
Cell-associated collagenolytic activity by Candida albicans.
PMID 11998872
Abstract
Cell associated collagenolytic activity of Candida albicans was quantified by measuring the degradation of synthetic peptide 2-furanacryloyl-Leu-Gly-Pro-Ala (FALGPA), which is a specific substrate for collagenase, by the freeze-thaw procedure method. This collagenolytic activity was enhanced by cells cultured in the presence of bovine serum albumin (BSA) in culture medium. However, this activity was inhibited in the presence of ethylenediaminetetraacetic acid disodium salt (EDTA-2Na), but not by the serine proteinase inhibitor p-amidinophenyl methanesulfonyl fluoride (APMSF), nor the aspartyl proteinase inhibitor pepstatin A. These results suggested the presence of a metalloenzyme on pericellular C. albicans.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F5135
N-[3-(2-Furyl)acryloyl]-Leu-Gly-Pro-Ala
C23H32N4O7