EMAIL THIS PAGE TO A FRIEND
Bioscience, biotechnology, and biochemistry
Kinetic resolutions of indan derivatives using bacteria.
PMID 11999429
Abstract
Racemic indan derivatives have been resolved by the hydrolysis of amide bonds using Corynebacterium ammoniagenes IFO12612 to produce (S)-amine and (R)-amides. In the kinetic resolution of 1 (N-12-(6-methoxy-indan-1-yl)ethyl]acetamide), it was possible to run the reaction to 44% conversion on a 10-g scale, obtaining (S)-amine 4 ((S)-2-(6-methoxy-indan-1-yl)ethylamine) at >99% enantiomeric excess (ee) and (R)-1 at 98% ee.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45789
Indan, analytical standard
C9H10
I1804
Indan, 95%
C9H10