EMAIL THIS PAGE TO A FRIEND
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry
Ethylammonium-selective membrane electrode using p-tert-butylcalix[6]arene derivatives.
PMID 11999526
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
83757
Cesium ionophore II, Selectophore, function tested, ≥97.0%
C90H120O18