EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Dissipation of foliar residues of diafenthiuron and its metabolites.
PMID 12012060
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
31571
Diafenthiuron, PESTANAL®, analytical standard
C23H32N2OS