EMAIL THIS PAGE TO A FRIEND
Inflammation research : official journal of the European Histamine Research Society ... [et al.]
Can urinary N-tele methylimidazoleacetic acid (t-MeImAA) serve as a marker of histaminergic activity in hepatic encephalopathy (HE)?
PMID 12013413