EMAIL THIS PAGE TO A FRIEND
Lancet (London, England)
Drug allergy transmitted by passionate kissing.
PMID 12020564
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B0070000 Bacampicillin hydrochloride, European Pharmacopoeia (EP) Reference Standard
C21H27N3O7S · HCl