EMAIL THIS PAGE TO A FRIEND
Toxicology in vitro : an international journal published in association with BIBRA
Modulation of hepatocyte thiol content by medium composition: implications for toxicity studies.
PMID 12020599
Abstract
Toxicity of compounds requiring glutathione for detoxification, thiol content and synthesis were determined in 24-h rat hepatocytes cultured in medium containing different concentrations of the sulphur amino acids. Glutathione synthesis was determined following prior depletion of glutathione with diethylmaleate. L-15 medium, which has high levels of cysteine and methionine (1 mM of each), provided some protection against dichloroacetone, dibromopropanol and dichloropropanol toxicity, and had a small effect on increasing glutathione content and synthesis, relative to Williams' medium E (WE) which has low levels (less than 0.5 mM) of both amino acids. However, WE containing N-acetylcysteine (NAC) (1 mM final cysteine concentration), with or without methionine (final concentration 1 mM), was a better cytoprotectant medium than L-15, markedly reducing toxicity of all three compounds, and rapidly (within 1.5 h) increasing cellular glutathione content. WE supplemented with methionine alone stimulated glutathione synthesis after an initial lag phase, and protected cultures against dichloropropanol, but not dibromopropanol or dichloroacetone, both of which are highly reactive in these cultures. There was a clear association between glutathione content at early time points in culture and toxicity observed at later time points, and overall these results indicate that differences in culture medium composition can alter intracellular glutathione content and xenobiotic toxicity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
372404
1,3-Dibromo-2-propanol, technical grade, 95%
C3H6Br2O
168548
1,3-Dichloroacetone, ≥95%
C3H4Cl2O