EMAIL THIS PAGE TO A FRIEND
The Journal of pharmacology and experimental therapeutics
SSR180575 (7-chloro-N,N,5-trimethyl-4-oxo-3-phenyl-3,5-dihydro-4H-pyridazino[4,5-b]indole-1-acetamide), a peripheral benzodiazepine receptor ligand, promotes neuronal survival and repair.
PMID 12023539
Abstract
In the present study, we have investigated the potential neuroprotective effects of a novel peripheral benzodiazepine binding site (PBR) ligand, 7-chloro-N,N,5-trimethyl-4-oxo-3-phenyl-3,5-dihydro-4H-pyridazino[4,5-b]indole-1-acetamide (SSR180575), in models of central and peripheral neurodegeneration in vivo and its effect on steroid concentrations in plasma and nervous tissue. SSR180575 shows high affinity (IC(50), 2.5-3.5 nM) and selectivity for the rat and human PBR and potently inhibits the in vivo binding of [(3)H]alpidem to PBR in the rat brain and spleen after oral or i.p. administration (ID(50), 0.1-0.3 mg/kg). In an experimental model of motoneuron degeneration induced by facial nerve axotomy in the immature rat, SSR180575 given i.p. or orally for 8 days rescued facial motoneurons, increasing their survival by 40 to 72% at 6 and 10 mg/kg p.o. b.i.d. Moreover, in this model, SSR180575 (10 mg/kg p.o. b.i.d.) increased by 87% the number of motoneurons immunoreactive to peripherin, a type III intermediate filament, whose expression is up-regulated during nerve regeneration. SSR180575 also improved functional recovery in acrylamide-induced neuropathy in the rat when given therapeutically at 2.5 to 10 mg/kg/day p.o. Furthermore, SSR180575 (3 mg/kg i.p. b.i.d.) accelerated functional recovery of the blink reflex after local injury of the facial nerve in the rat. SSR180575 increased pregnenolone accumulation in the brain and sciatic nerve (+100% at 3 mg/kg i.p.), suggesting that its neuroprotective effects are steroid-mediated. These results indicate that PBR ligands (e.g., SSR180575) promote neuronal survival and repair in axotomy and neuropathy models and have potential for the treatment of neurodegenerative diseases (e.g., peripheral neuropathies or amyotrophic lateral sclerosis).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A1862
Alpidem, ≥98% (HPLC), powder
C21H23Cl2N3O