EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Endothelin-1 induces vascular endothelial growth factor by increasing hypoxia-inducible factor-1alpha in ovarian carcinoma cells.
PMID 12023962
Abstract
Angiogenesis is an essential prerequisite for tumor growth, invasion, and metastasis. In ovarian carcinoma cells, endothelin-1 (ET-1) stimulates the secretion of vascular endothelial growth factor (VEGF), a major mediator of tumor angiogenesis. In OVCA 433 and HEY ovarian carcinoma cell lines, ET-1 treatment increases VEGF mRNA expression and induces VEGF protein levels in a time- and dose-dependent fashion, and do so to a greater extent under hypoxic conditions. ET-1 also increases hypoxia-inducible factor-1alpha (HIF-1alpha) accumulation and activates the HIF-1 transcription complex under both normoxic and hypoxic conditions, suggesting a role for HIF-1 in the induction of VEGF expression. These effects are inhibited by the selective ET(A) receptor (ET(A)R) antagonist, BQ123. The ET-1-induced increase in HIF-1alpha protein levels is due to the enhanced HIF-1alpha stabilization. These results implicate HIF-1alpha in the induction of VEGF expression in ET-1-stimulated ovarian carcinoma cells, and provide a mechanism whereby ET-1 acting selectively through ET(A)R can interact with the HIF-1alpha-dependent machinery of angiogenesis. Our results suggest that new therapeutic strategies using specific ET(A)R antagonists could provide an additional approach to the treatment of ovarian carcinoma by inhibiting neovascularization as well as tumor cell growth.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E7764
Endothelin 1, ≥97% (HPLC), powder
C109H159N25O32S5