EMAIL THIS PAGE TO A FRIEND
Drug and chemical toxicology
Initial study in rats evaluating the effects of 1,4-dichlorobutene-2 (DCB) on the respiratory tract.
PMID 12024805
Abstract
Rats were exposed by inhalation to either 0.5 ppm 1,4-dichlorobutene-2 (DCB) for two years or to 5.0 ppm for seven months, 2.5 ppm for five months, and no further exposure for 12 months prior to sacrifice. Malignant and non-malignant tumors of the nasal tissues were seen in both test groups with the incidence and proportion of malignant tumors being much higher in the 5.0/2.5 ppm rats. Under the conditions of this study, DCB is carcinogenic in rats of both sexes.