EMAIL THIS PAGE TO A FRIEND
The practising midwife
Lotions and lubricants.
PMID 12026657
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A8986
Alexidine dihydrochloride, ≥95% (HPLC)
C26H56N10 · 2HCl