EMAIL THIS PAGE TO A FRIEND
Journal of the American Geriatrics Society
Incorrect assumption regarding the mechanism of action of flavoxate.
PMID 12028232
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F8304
Flavoxate hydrochloride, ≥98% (HPLC), solid
C24H25NO4 · HCl