EMAIL THIS PAGE TO A FRIEND
Journal of oral rehabilitation
Accuracy of check-bite registration and centric condylar position.
PMID 12028494
Abstract
In a dentate subject a jaw relation can either be determined in maximum intercuspation and is as such given by the occlusal morphology, or the mandibular position can be allocated according to the centric position of the condyles. For comprehensive restorative treatment or analytic measures of the occlusion it is important to record the centric condylar position. Various registration methods have been described in the literature, but there is no consensus on which is the 'best'. The aim of the present study was therefore to assess the accuracy of various registration methods and evaluate a possible influence of the used materials. Four dentists were involved in the clinical part of the study, another was responsible for the measurements. Impressions were taken from 81 fully dentate volunteers. The casts were mounted by face-bow transfer and central-bearing-point (CBP) registration into Dentatus articulators. Subsequently the centric condylar position was determined with six different methods and materials, respectively. Each method was reproduced twice so that a total of 18 registrations was performed per patient. The mandibular positions which resulted from the individual registrations were then repeatedly compared in the condylar area using a computer supported specially modified measuring articulator. The accuracy was found best for the unrefined wax wafer registration (x=0.33 mm) and with an average of 0.44 mm worst when using acrylic wafers. The CBP and frontal jig methods as well as tin-foil and refined wax wafers showed an accuracy in-between these boundaries. The biggest measured mandibular displacement between any two registrations were considerably 2.0 mm. However, the described differences in accuracy between the various methods and materials proved statistically not significant. All investigated jaw registrations showed an accuracy of about 20 times the tactile fine sensibility of natural teeth which has to be taken into account when inserting fixed prosthetic restorations in centric condylar position. Despite meticulous clinical and technical procedures small occlusal adjustments are therefore almost unavoidable.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF26583194
Tin, foil, 150x150mm, thickness 0.125mm, as rolled, 98.8%
Sn
GF66339254
Tin, foil, 150x150mm, thickness 0.125mm, as rolled, 99.99+%
Sn
GF54760259
Tin, foil, 150x150mm, thickness 0.15mm, as rolled, 98.8%
Sn
GF67327375
Tin, foil, 150x150mm, thickness 0.1mm, as rolled, 98.8%
Sn
GF27720019
Tin, foil, 150x150mm, thickness 0.1mm, as rolled, 99.95%
Sn
GF16685219
Tin, foil, 150x150mm, thickness 0.20mm, as rolled, 98.8%
Sn
GF45567434
Tin, foil, 150x150mm, thickness 0.25mm, as rolled, 99.99+%
Sn
GF28603212
Tin, foil, 150x150mm, thickness 0.5mm, as rolled, 98.8%
Sn
GF28221274
Tin, foil, 150x150mm, thickness 0.5mm, as rolled, 99.99+%
Sn
GF77851323
Tin, foil, 150x150mm, thickness 1.0mm, as rolled, 98.8%
Sn
GF16505636
Tin, foil, 150x150mm, thickness 1.0mm, as rolled, 99.999%
Sn
GF50720925
Tin, foil, 150x150mm, thickness 2.0mm, as rolled, 98.8%
Sn
GF00846413
Tin, foil, 150x150mm, thickness 2.0mm, as rolled, 99.99+%
Sn
GF99151881
Tin, foil, 25x25mm, thickness 0.125mm, as rolled, 98.8%
Sn
GF83193925
Tin, foil, 25x25mm, thickness 0.125mm, as rolled, 99.99+%
Sn
GF86850029
Tin, foil, 25x25mm, thickness 0.15mm, as rolled, 98.8%
Sn
GF17353425
Tin, foil, 25x25mm, thickness 0.15mm, as rolled, 99.99+%
Sn
GF40941097
Tin, foil, 25x25mm, thickness 0.1mm, as rolled, 98.8%
Sn
GF66628996
Tin, foil, 4mm disks, thickness 0.007mm, 97.4%
Sn
GF33080532
Tin, foil, 4mm disks, thickness 0.015mm, 99.75%
Sn
GF29177039
Tin, foil, 4mm disks, thickness 0.02mm, 99.75%
Sn
GF51371232
Tin, foil, 4mm disks, thickness 0.125mm, as rolled, 98.8%
Sn
GF34214464
Tin, foil, 4mm disks, thickness 0.25mm, as rolled, 98.8%
Sn
GF63204106
Tin, foil, 4mm disks, thickness 1.0mm, as rolled, 100%
Sn
GF04830331
Tin, foil, 50mm disks, thickness 0.0125mm, 97.4%
Sn
GF96864643
Tin, foil, 50mm disks, thickness 0.05mm, as rolled, 98.8%
Sn
GF07913088
Tin, foil, 8mm disks, thickness 0.06mm, as rolled, 99.99+%
Sn
GF18183764
Tin, foil, 8mm disks, thickness 0.1mm, as rolled, 98.8%
Sn
GF36451098
Tin, foil, 8mm disks, thickness 0.65mm, as rolled, 99.99+%
Sn
GF00022699
Tin, foil, light tested, 100x100mm, thickness 0.0125mm, 97.4%
Sn