EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Electrochemical and peroxidase oxidation study of N'-hydroxyguanidine derivatives as NO donors.
PMID 12031330
Abstract
The electrochemical properties of a series of N-substituted-N'-hydroxyguanidines were studied. Two oxidation potentials of each compound were obtained by cyclic voltammetry. The E(ox1) values were from 0.51 to 0.62V, while the E(ox2) values were from 1.14 to 1.81V in acetonitrile solution. Next, their enzymatic controlled NO release abilities were evaluated. All N'-hydroxyguanidines exhibited efficient NO release abilities under the oxidation by horseradish peroxidase in the presence of H(2)O(2).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H7654
Hydroxyguanidine sulfate salt
CH5N3O · 0.5H2O4S