EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Inhibition of glutathione S-transferase in rat hepatocytes by a glycine-tetrazole modified S-alkyl-GSH analogue.
PMID 12039566
Abstract
Glutathione (GSH) conjugates inhibit enzymes that are involved in drug metabolism and drug resistance, but their cellular uptake is very low. To improve membrane-permeability, we synthesized a novel GSH-conjugate analogue with a tetrazole carboxylate isostere at the glycine position. Introduction of the tetrazole decreases inhibitory potency towards CDNB conjugation by glutathione S-transferase. However, the tetrazole derivative inhibited 2-bromoisovalerylurea conjugation in rat liver cytosol, as well as in hepatocytes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
292389
2-Bromoheptane, technical grade
C7H15Br