EMAIL THIS PAGE TO A FRIEND
European biophysics journal : EBJ
Amiloride derivatives block ion channel activity and enhancement of virus-like particle budding caused by HIV-1 protein Vpu.
PMID 12046895
Abstract
The Vpu protein of human immunodeficiency virus type 1 forms cation-selective ion channels and enhances the process of virion budding and release. Mutagenesis studies have shown that the N-terminal transmembrane domain primarily controls both of these activities. Here we report that the Vpu ion channel is inhibited by the amiloride derivatives 5-(N,N-hexamethylene)amiloride and 5-(N,N-dimethyl)amiloride but not by amiloride itself, nor by amantadine. Hexamethyleneamiloride also inhibits budding of virus-like particles from HeLa cells expressing HIV-1 Gag and Vpu proteins. These results confirm the link between Vpu ion channel activity and the budding process and also suggest that amiloride derivatives might have useful anti-HIV-1 properties.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A9561
5-(N,N-Hexamethylene)amiloride
C12H18ClN7O