EMAIL THIS PAGE TO A FRIEND
International journal of toxicology
The blockade of mineralocorticoid hormone signaling provokes dramatic teratogenesis in cultured rat embryos.
PMID 12055020
Abstract
Although the administration of adrenocortical hormones to pregnant rats provokes only limited effect on the growth and development of the fetus, the direct influence of these steroids on cultured embryos has never been studied. The disruption of cell signaling by ZK 91587, which specifically occupies the mineralocorticoid receptor, resulted within 2 days in significant and pronounced adverse effects on the total length, the somite number, the embryo curvature, the communication between vitelline and umbilical blood vessels in the allantoid, and the vascularization of the vitelline sac, in 244-hour Wistar rat embryos in culture. The average score of 16 organs declined in a dose-dependent manner, following exposure to ZK 91587, and this was totally reversed by 10 microM aldosterone which, by itself, did not at all influence the embryonic development. The organogenesis was inhibited in the order: hind limb > fore limb > optic stalk > brain > olfactory pit > otic vesicle. ZK 91587 was completely ineffective in embryos that had attained the age of 260 hours. Similar, but less dramatic, results were obtained with the mineralocorticoid antagonist RU 26752, and with the antiglucocorticoid RU 38486. Sprague-Dawley rat embryos responded in a manner similar to the Wistar conceptuses. Thus, steroid receptor-mediated cell signaling is of critical importance to the growth and development of cultured rat embryos, which form a new model system to unravel adrenocortical hormone action.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R2153
RU 26752, ≥98% (HPLC), solid
C25H36O3