EMAIL THIS PAGE TO A FRIEND
Biochemistry
The role of surface-exposed lysines in wrapping DNA about the bacterial histone-like protein HU.
PMID 12056890
Abstract
Several basic proteins, including the ubiquitous HU proteins, serve histone-like functions in prokaryotes. Significant sequence conservation exists between HU homologues; yet binding sites varying from 9 to 37 bp have been reported. TF1, an HU homologue with a 37 bp binding site that is encoded by the Bacillus subtilis bacteriophage SPO1, binds with nM affinity to DNA that contains 5-hydroxymethyluracil (hmU) in place of thymine and to T-containing DNA with loops. We evaluated the contribution of three conserved lysines to specifying the length of the binding site and show that Lys3 is critical for maintaining a long binding site in T-containing DNA: A mutant protein in which Lys3 is replaced with Gln(TF1-K3Q) is completely deficient in forming a stable complex. The affinity for 37 bp hmU-containing DNA is also reduced, from approximately 3 nM for wild-type TF1 to approximately 90 nM for TF1-K3Q. The decrease in affinity of TF1-K3Q for hmU-containing DNA > or = 25 bp suggests that Lys3 contacts DNA 8-9 bp distal to the sites of kinking. We propose that Lys3 forms an internal saltbridge to Asp26 in HU homologues characterized by shorter binding sites and that its surface exposure, and hence a longer binding site, may correlate with absence of this aspartate.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
852589
5-(Hydroxymethyl)uracil, 97%
C5H6N2O3