EMAIL THIS PAGE TO A FRIEND
International journal for parasitology
Evidence for de novo sphingolipid biosynthesis in Toxoplasma gondii.
PMID 12062486
Abstract
Glycolipids are important components of cellular membranes involved in various biological functions. In this report, we describe the identification of the de novo synthesis of glycosphingolipids by Toxoplasma gondii tachyzoites. Parasite-specific glycolipids were identified by metabolic labelling of parasites with tritiated serine and galactose. These glycolipids were characterised as sphingolipids based on the labelling protocol and their insensitivity towards alkaline treatment. Synthesis of parasite glycosphingolipids were inhibited by threo-phenyl-2-palmitoylamino-3-morpholino-1-propanol and L-cycloserine, two well-established inhibitors of de novo sphingolipid biosynthesis. The identified glycolipids were insensitive towards treatment with endoglycoceramidase II indicating that they might belong to globo-type glycosphingolipids. Taken together, we provide evidence for the first time that T. gondii is capable of synthesising glycosphingolipids de novo.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E9030 Endoglycoceramidase II from Rhodococcus sp., aqueous solution