EMAIL THIS PAGE TO A FRIEND
Toxicology
Determination of the sensitising activity of the rubber contact sensitisers TMTD, ZDMC, MBT and DEA in a modified local lymph node assay and the effect of sodium dodecyl sulfate pretreatment on local lymph node responses.
PMID 12062936
Abstract
A modified local lymph node assay (LLNA) was used to determine the sensitising activity of four chemicals used for the production of natural rubber latex products. Tetramethylthiuramdisulfide (TMTD), 2-mercaptobenzothiazole (MBT) and zincdimethyldithiocarbamate (ZDMC), three moderate human sensitisers, and diethylamine (DEA) a known human sensitiser, were epicutaneously administered on the ear and the proliferating activity in the draining (auricular) lymph node (LN) was determined by ex vivo (3)H-thymidine incorporation. Consistent results were obtained for TMTD and ZDMC with stimulation indices (SI) above 3, identifying these compounds as sensitiser, while for DEA and MBT inconsistent results were obtained. For all parameters determined such as LN weight, LN cell number, cell proliferation per 2 x 10(6) cells, and cell proliferation per LN statistical significant increases were observed. The SI, expressed as cellular proliferation per LN or per animal (left and right LN combined), was the most sensitive parameter with an optimum at day 5 after start of treatment.Furthermore, we investigated whether the use of sodium dodecyl sulfate (SDS) was able to enhance weak responses in the LLNA. SDS treatment with dosages of 10% and higher resulted in a SI above 3, while a dosage of 1% SDS showed no activity. Pretreatment with 1% SDS 1 h before application of the rubber chemicals enhanced the responses to these chemicals consistently, identifying also DEA and MBT as sensitisers. Our results indicate that SDS had synergistic activity on the LN responses of the administered rubber chemicals in the LLNA. For the moderately responding sensitisers TMTD and ZDMC both IFN-gamma and IL-4 production was observed. For the weakly responding sensitisers DEA and MBT both IFN-gamma and IL-4 cytokine production was only observed after pretreatment of the animals with 10% SDS. For 10% and 20% SDS, inducing approximately a SI of 20 in the LLNA, no induction of cytokines was observed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
329711
Zinc dimethyldithiocarbamate, 97%
C6H12N2S4Zn
45708
Ziram, PESTANAL®, analytical standard
C6H12N2S4Zn