EMAIL THIS PAGE TO A FRIEND
Orvosi hetilap
[Cross-reaction potentials of ketoprofen].
PMID 12063858
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T1410900 Tiaprofenic acid, European Pharmacopoeia (EP) Reference Standard
C14H12O3S