EMAIL THIS PAGE TO A FRIEND
Health technology assessment (Winchester, England)
Zanamivir for the treatment of influenza in adults: a systematic review and economic evaluation.
PMID 12065066
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0492
Zanamivir, ≥98% (HPLC)
C12H20N4O7