EMAIL THIS PAGE TO A FRIEND
Pharmacology & toxicology
Cytochrome P450 enzymes contributing to demethylation of maprotiline in man.
PMID 12071336
Abstract
From case reports of patients treated with the tetracyclic antidepressant drug maprotiline, it appears that this drug is subject to polymorphic metabolism. Thus, we studied formation of the major maprotiline metabolite desmethylmaprotiline to identify the human cytochrome P-450 enzymes (CYP) involved. In incubations with human liver microsomes from two different donors, the substrate maprotiline was used at five different concentrations (5 to 500 microM). For selective inhibition of CYPs, quinidine (0.5-50 microM; CYP2D6), furafylline (0.3-30 microM; CYP1A2), ketoconazole (0.2-20 microM; CYP3A4), mephenytoin (20-200 microM; CYP2C19), chlorzoxazone (1-100 microM; CYP2E1), sulphaphenazole (0.2-100 microM; CYP2C9) and coumarin (0.2-100 microM; CYP2A6) were used. Desmethylmaprotiline concentrations were measured by HPLC, and enzyme kinetic parameters were estimated using extended Michaelis-Menten equations with non-linear regression. Relevant inhibition of the desmethylmaprotiline formation rate was observed in incubations with quinidine, furafylline and ketoconazole only. Formation rates of desmethylmaprotiline were consistent with a two enzyme model with a high (K(M)=71 and 84 microM) and a low (K(M)=531 and 426 microM) affinity site for maprotiline in the two samples, respectively. The high affinity site was competitively inhibited by quinidine (K(i,nc) 0.13 and 0.61 microM), the low-affinity site was non-competitively inhibited by furafylline (K(i,nc) 0.11 and 1.3 microM). Thus it appears that CYP2D6 and CYPIA2 contribute to maprotiline demethylation. Based on the parameters obtained, for plasma concentrations of 1 microM 83% (mean) of desmethylmaprotiline formation in vivo is expected to be mediated by CYP2D6 while 17% only may be attributed to CYPIA2 activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F124
Furafylline, ≥98% (HPLC)
C12H12N4O3