EMAIL THIS PAGE TO A FRIEND
Contact dermatitis
Ammonium thiolactate and thiolactic acid: important hairdressers' allergens?
PMID 12081707
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W318000
2-Mercaptopropionic acid, ≥95%, FG
C3H6O2S
T31003
Thiolactic acid, 95%
C3H6O2S