EMAIL THIS PAGE TO A FRIEND
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry
Crystal structure of 3-amino-1,2,4-triazin-5(2H)-one.
PMID 12083566
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
100625
3-Amino-1,2,4-triazine, 97%
C3H4N4